Supelco® Drug and metabolite standards, nalbuphine hydrochloride solution, (CRM)
N-051-1ML
: Merck
:
- Koncentrace:1,0 mg/ml in methanol (as free base)
- Bal.:1 mL
- Standard:Certified reference material
- Čí. CAS:23277-43-2
- Popis:Nalbuphine hydrochloride solution
- Popis:Nalbuphine hydrochloride solution
- Pk Info:ampoule of 1 ml