Supelco® Drug and metabolite standards, Ibuprofen solution, (CRM)
Catalog # I-009-1ML
Supplier: Merck
CAS Number:
Specifications
- Koncentrace:1,0 mg/ml in methanol
- Bal.:1 mL
- Standard:Certified reference material
- Čí. CAS:15687-27-1
- Popis:Ibuprofen solution
- Popis:Ibuprofen solution
- Pk Info:ampoule of 1 ml