Drug and metabolite standards, EMDP hydrochloride solution, (CRM), Supelco®
Catalog # E-057-1ML
Supplier: Merck
CAS Number:
Specifications
- Koncentrace:1,0 mg/ml in methanol (as free base)
- Bal.:1 mL
- Standard:Certified reference material
- Čí. CAS:102177-19-5
- Popis:EMDP hydrochloride solution
- Popis:EMDP hydrochloride solution
- Pk Info:ampoule of 1 ml