Drug and metabolite standards, EDDP perchlorate solution, (CRM), Supelco®
Catalog # E-022-1ML
Supplier: Merck
CAS Number:
Specifications
- Koncentrace:1,0 mg/ml in methanol (as pyrolinium)
- Bal.:1 mL
- Standard:Certified reference material
- Čí. CAS:66729-78-0
- Popis:EDDP perchlorate solution
- Popis:EDDP perchlorate solution
- Pk Info:ampoule of 1 ml