Supelco® Drug and metabolite standards, Dextromethorphan solution, (CRM)
Catalog # D-013-1ML
Supplier: Merck
CAS Number:
Specifications
- Koncentrace:1,0 mg/ml in methanol
- Bal.:1 mL
- Standard:Certified reference material
- Čí. CAS:125-71-3
- Popis:Dextromethorphan solution
- Popis:Dextromethorphan solution
- Pk Info:ampoule of 1 ml