Supelco® Cerilliant® Organic Reference Standard, Catecholamine Metabolites Mix solution
C-111-1ML
: Merck
:
- Koncentrace:1 mg/ml each component in methanol
- Bal.:1 mL
- Standard:Certified reference material, Cerilliant®
- Popis:Catecholamine Metabolites mix solution
- Podmínky uskladnění:–70 °C
- Popis rizikové třídy:H225 - H301 + H311 + H331 - H370
- Podmínky k odeslání:WICE
- M-Clarity quality level:MQ300
- Molekulární hmotnost:198,17
- Bod vzplanutí:9,7 °C - Closed cup
- Pk Info:Ampule of 1 ml
- GHS signal: GHS02, GHS06, GHS08
: P210 - P280 - P301 + P310 + P330 - P302 + P352 + P312 - P304 + P340 + P311