Supelco® Drug and metabolite standards, Acetaminophen solution, (CRM)
Catalog # A-064-1ML
Supplier: Merck
CAS Number:
Specifications
- Koncentrace:1,0 mg/ml in methanol
- Bal.:1 mL
- Standard:Certified reference material
- Čí. CAS:103-90-2
- Popis:Acetaminophen solution
- Popis:Acetaminophen solution
- Pk Info:ampoule of 1 ml