Order Entry
Czech Republic
ContactUsLinkComponent
Drug and metabolite standards, 6beta-Naltrexol solution, (CRM), Supelco®
Drug and metabolite standards, 6beta-Naltrexol solution, (CRM), Supelco®
Catalog # N-038-1ML
Supplier:  Merck
CAS Number:  
undefined
Drug and metabolite standards, 6beta-Naltrexol solution, (CRM), Supelco®
Catalog # N-038-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Koncentrace:
    1,0 mg/ml in methanol
  • Bal.:
    1 mL
  • Standard:
    Certified reference material
  • Čí. CAS:
    49625-89-0
  • Popis:
    6beta-Naltrexol solution
  • Popis:
    6beta-Naltrexol solution
  • Pk Info:
    ampoule of 1 ml

Specifications

About this item