Supelco® Organic Reference Standard, Linolenic Acid Methyl Ester Isomer Mix
CRM47792
: Merck
:
- Koncentrace:10 mg/ml in dichloromethane
- Bal.:1 Ampoul
- Standard:Certified reference material
- Popis:Linolenic Acid methyl ester isomer mix
- Podmínky uskladnění:–10...–25 °C
- Podmínky k odeslání:WICE
- M-Clarity quality level:MQ100
- Bod vzplanutí:100 °C
- Pk Info:Ampule of 1 ml
- GHS signal: GHS08, GHS07