HPLC Columns, OPTIMAPACK
: HICHROM
OPTIMAPAK is a range of HPLC columns which are available in three bonded phases and an unbonded silica phase.
- Surface area: 330 m2/g
- Catalog No:
- HISMOPSIL-51002546
- HISMOP-51002546
- Naplnění uhlíkem:
- —
- 20%
- S navazanou skupinou:
- Yes
- Yes
- Povrchová oblast:
- 330 m2/g
- 330 m2/g
Frequently Bought Together