0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Filtry
Patrony filtrační
Accessories for cartridges for deionisers, arium®
Accessories for cartridges for deionisers, arium®
VIVA615DIC01
:
Sartorius
Accessories for cartridges for deionisers, arium®
VIVA615DIC01
:
Sartorius
Popis:
Ion exchange resin mix, refill pack for complete deionisation cartridges, arium® 615DI, in 25 l PE bag
Bal.:
1