0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Filtry
Patrony filtrační
Cartridge for deionisers, arium®
Cartridge for deionisers, arium®
VIVA615DI014
:
Sartorius
Cartridge for deionisers, arium®
VIVA615DI014
:
Sartorius
Popis:
Deionisation cartridge with installation kit, capacity range up to 1400 l @ 20 ºdH, flow rate 600 l/h
Bal.:
1