Safety helmets, uvex thermo boss
Catalog # UVEX9754.300
Supplier: UVEX
Specifications
- Barva:Červený
- Popis:Safety helmet with slots, uvex thermo boss
- Bal.:1
Specifications
About this item
This uvex thermo boss helmet features a six-point textile band attachment which ensures an optimal fit and high comfort and a conventional suspension harness for comfortable adjustable fit.
- Made from heat-resistant polycarbonate
- Side Euroslot adapters
Suitable for use in high ambient temperatures.
Certifications: EN 397, high temperatures (150 °C)