Screw cap with septum for microcentrifuge tubes, Micrewtube™
Catalog # SIMPT347AQX
Supplier: Simport Scientific
Specifications
- Popis:Septum screw cap, aqua, for Micrew™ microcentrifuge tubes
- Materiál:PP
- Bal.:250
Specifications
About this item
This screw cap incorporates a pierceable septum made of chemically resistant PTFE on the outside and silicone on the inside, both components being stable over a broad range of temperatures. The septum also acts as a silicone o-ring for better sample protection.
- Made of polypropylene
- Autoclavable
It is especially made to fit and be used with all Micrew™ microcentrifuge tubes. The cap is pierceable with pipette tips as well as with syringe needles.
Certifications: Microcentrifuge tubes and caps are certified RNase, DNase, DNA and Pyrogen free.