Specific Gravity Hydrometers
Dodavatel: Amarell
O tomto produktu
These general-purpose hydrometers are available with a thermometer inside the body or without a thermometer.
- Capillary prismatic
- Reliable and simple
- Paper scale with type designation
- Shot weighted body
: Supplied in a transparent plastic case.
53 Dostupných variant níže
Product Details & Documents
These general-purpose hydrometers are available with a thermometer inside the body or without a thermometer.
- Capillary prismatic
- Reliable and simple
- Paper scale with type designation
- Shot weighted body
: Supplied in a transparent plastic case.
Specifications for Product Family
Specifikace
- Catalog No:
- AMARH826760-FL
- AMARH826764-FL
- AMARH826780
- AMARH826782
- AMARH826784
- AMARH826786
- AMARH826788
- AMARH826790
- AMARH826792
- AMARH826910
- AMARH826912
- AMARH826914
- AMARH826916
- AMARH826918
- AMARH826920
- AMARH826922
- AMARH826924
- AMARH826926
- AMARH826928
- AMARH826930
- AMARH826932
- AMARH826934
- AMARH826936
- AMARH826938
- AMARH826940
- AMARH826942
- AMARH826500
- AMARH826502
- AMARH826504
- AMARH826506
- AMARH826508
- AMARH826510
- AMARH826512
- AMARH826628
- AMARH826514
- AMARH826630
- AMARH826516
- AMARH826632
- AMARH826634
- AMARH826636
- AMARH826638
- AMARH826640
- AMARH826642
- AMARH826644
- AMARH826646
- AMARH826648
- AMARH826786DKD3
- AMARH826788DKD3
- AMARH826790DKD3
- AMARH826640/DKD1
- AMARH826786WC
- AMARH826640/DKD3
- AMARH826788WC
- Přesnost:
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- ±1 scale division
- Referenční teplota:
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
- 60/60 °F (except model AMARH826764-FL: 15 °C)
Specifikace
Product Family Options
With thermometer
Informace o produktu
- SekvenceModelRozsahDěleníTeplotní rozsahDélkaDostupnostPrice
- AMARH826760-FLSpecific gravity hydrometer, filling: with red or blue special liquidSpecific gravity hydrometer, filling: with red or blue special liquid01101 H0,500 - 0,650 °SG0,001 °SG+30...+90 °F; –1,1...+32,2 °C360 mmPřihlásit se pro dostupnost1 * 1 KS4 840,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826760-FLPřesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826764-FLSpecific gravity hydrometer, filling: with red or blue special liquidSpecific gravity hydrometer, filling: with red or blue special liquid02310 H500 - 650 °SG0,001 °SG0...0,5 °C; 35...0,5 °C390 mmPřihlásit se pro dostupnost1 * 1 KS5 280,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826764-FLPřesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C)
Without thermometer
Informace o produktu
- SekvenceModelRozsahDěleníTeplotní rozsahDélkaDostupnostPrice
- AMARH826780Specific gravity hydrometerSpecific gravity hydrometer03102 H - 620,650 - 0,700 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 150,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826780Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826782Specific gravity hydrometerSpecific gravity hydrometer04103 H - 620,700 - 0,750 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 150,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826782Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826784Specific gravity hydrometerSpecific gravity hydrometer05104 H - 620,750 - 0,800 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 150,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826784Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826786Specific gravity hydrometerSpecific gravity hydrometer06105 H - 620,800 - 0,850 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 150,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826786Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826788Specific gravity hydrometerSpecific gravity hydrometer07106 H - 620,850 - 0,900 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 150,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826788Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826790Specific gravity hydrometerSpecific gravity hydrometer08107 H - 620,900 - 0,950 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 150,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826790Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826792Specific gravity hydrometerSpecific gravity hydrometer09108 H - 620,950 - 1,000 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 150,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826792Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826910Specific gravity hydrometerSpecific gravity hydrometer10125 H - 621,000 - 1,050 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 190,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826910Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826912Specific gravity hydrometerSpecific gravity hydrometer11126 H - 621,050 - 1,100 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 190,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826912Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826914Specific gravity hydrometerSpecific gravity hydrometer12127 H - 621,100 - 1,150 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 190,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826914Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826916Specific gravity hydrometerSpecific gravity hydrometer13128 H - 621,150 - 1,200 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 190,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826916Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826918Specific gravity hydrometerSpecific gravity hydrometer14129 H - 621,200 - 1,250 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 190,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826918Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826920Specific gravity hydrometerSpecific gravity hydrometer15130 H - 621,250 - 1,300 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 190,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826920Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826922Specific gravity hydrometerSpecific gravity hydrometer16131 H - 621,300 - 1,350 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 360,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826922Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826924Specific gravity hydrometerSpecific gravity hydrometer17132 H - 621,350 - 1,400 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 360,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826924Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826926Specific gravity hydrometerSpecific gravity hydrometer18133 H - 621,400 - 1,450 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 360,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826926Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826928Specific gravity hydrometerSpecific gravity hydrometer19134 H - 621,450 - 1,500 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 360,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826928Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826930Specific gravity hydrometerSpecific gravity hydrometer20135 H - 621,500 - 1,550 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 360,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826930Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826932Specific gravity hydrometerSpecific gravity hydrometer21136 H - 621,550 - 1,600 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 360,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826932Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826934Specific gravity hydrometerSpecific gravity hydrometer22137 H - 621,600 - 1,650 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 580,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826934Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826936Specific gravity hydrometerSpecific gravity hydrometer23138 H - 621,650 - 1,700 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 580,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826936Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826938Specific gravity hydrometerSpecific gravity hydrometer24139 H - 621,700 - 1,750 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 580,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826938Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826940Specific gravity hydrometerSpecific gravity hydrometer25140 H - 621,750 - 1,800 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 580,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826940Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826942Specific gravity hydrometerSpecific gravity hydrometer26141 H - 621,800 - 1,850 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 580,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826942Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826500Specific gravity hydrometerSpecific gravity hydrometer2782 H - 620,650 - 0,700 °SG0,005 °SG330 mmPřihlásit se pro dostupnost1 * 1 KS1 480,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826500Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826502Specific gravity hydrometerSpecific gravity hydrometer2883 H - 620,700 - 0,750 °SG0,005 °SG330 mmPřihlásit se pro dostupnost1 * 1 KS1 480,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826502Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826504Specific gravity hydrometerSpecific gravity hydrometer2984 H - 620,750 - 0,800 °SG0,005 °SG330 mmPřihlásit se pro dostupnost1 * 1 KS1 480,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826504Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826506Specific gravity hydrometerSpecific gravity hydrometer3085 H - 620,800 - 0,850 °SG0,005 °SG330 mmPřihlásit se pro dostupnost1 * 1 KS1 480,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826506Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826508Specific gravity hydrometerSpecific gravity hydrometer3186 H - 620,850 - 0,900 °SG0,005 °SG330 mmPřihlásit se pro dostupnost1 * 1 KS1 480,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826508Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826510Specific gravity hydrometerSpecific gravity hydrometer3287 H - 620,900 - 0,950 °SG0,005 °SG330 mmPřihlásit se pro dostupnost1 * 1 KS1 480,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826510Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826512Specific gravity hydrometerSpecific gravity hydrometer3388 H - 620,950 - 1,000 °SG0,005 °SG330 mmPřihlásit se pro dostupnost1 * 1 KS1 520,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826512Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826628Specific gravity hydrometerSpecific gravity hydrometer3498 H - 620,950 - 1,000 °SG0,005 °SG330 mmPřihlásit se pro dostupnost1 * 1 KS1 520,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826628Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826514Specific gravity hydrometerSpecific gravity hydrometer3589 H - 621,000 - 1,050 °SG0,005 °SG330 mmPřihlásit se pro dostupnost1 * 1 KS1 520,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826514Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826630Specific gravity hydrometerSpecific gravity hydrometer36111 H - 621,000 - 1,050 °SG0,005 °SG330 mmPřihlásit se pro dostupnost1 * 1 KS1 520,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826630Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826516Specific gravity hydrometerSpecific gravity hydrometer3790 H - 621,050 - 1,100 °SG0,005 °SG330 mmPřihlásit se pro dostupnost1 * 1 KS1 520,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826516Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826632Specific gravity hydrometerSpecific gravity hydrometer38112 H - 621,050 - 1,100 °SG0,005 °SG330 mmPřihlásit se pro dostupnost1 * 1 KS1 520,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826632Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826634Specific gravity hydrometerSpecific gravity hydrometer39113 H - 621,100 - 1,150 °SG0,005 °SG330 mmPřihlásit se pro dostupnost1 * 1 KS1 520,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826634Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826636Specific gravity hydrometerSpecific gravity hydrometer40114 H - 621,150 - 1,200 °SG0,005 °SG330 mmPřihlásit se pro dostupnost1 * 1 KS1 520,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826636Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826638Specific gravity hydrometerSpecific gravity hydrometer41115 H - 621,200 - 1,250 °SG0,005 °SG330 mmPřihlásit se pro dostupnost1 * 1 KS1 520,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826638Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826640Specific gravity hydrometerSpecific gravity hydrometer42116 H - 621,250 - 1,300 °SG0,005 °SG330 mmPřihlásit se pro dostupnost1 * 1 KS1 520,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826640Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826642Specific gravity hydrometerSpecific gravity hydrometer43117 H - 621,300 - 1,350 °SG0,005 °SG330 mmPřihlásit se pro dostupnost1 * 1 KS2 630,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826642Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826644Specific gravity hydrometerSpecific gravity hydrometer44118 H - 621,350 - 1,400 °SG0,005 °SG330 mmPřihlásit se pro dostupnost1 * 1 KS2 630,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826644Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826646Specific gravity hydrometerSpecific gravity hydrometer45119 H - 621,400 - 1,450 °SG0,005 °SG330 mmPřihlásit se pro dostupnost1 * 1 KS2 630,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826646Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826648Specific gravity hydrometerSpecific gravity hydrometer46120 H - 621,450 - 1,500 °SG0,005 °SG330 mmPřihlásit se pro dostupnost1 * 1 KS2 630,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826648Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C)
With DKD certificate
Informace o produktu
- SekvenceModelRozsahDěleníTeplotní rozsahDélkaDostupnostPrice
- AMARH826786DKD3Specific gravity hydrometerSpecific gravity hydrometer47105 H - 620,800 - 0,850 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KSSpecifikace:
More
Specifications
Cat. No.AMARH826786DKD3Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826788DKD3Specific gravity hydrometerSpecific gravity hydrometer48106 H - 620,850 - 0,900 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KSSpecifikace:
More
Specifications
Cat. No.AMARH826788DKD3Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826790DKD3Specific gravity hydrometerSpecific gravity hydrometer49107 H - 620,900 - 0,950 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KSSpecifikace:
More
Specifications
Cat. No.AMARH826790DKD3Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826640/DKD1Specific gravity hydrometerSpecific gravity hydrometer50116 H - 621,250 - 1,300 °SG0,005 °SG330 mmPřihlásit se pro dostupnost1 * 1 KSSpecifikace:
More
Specifications
Cat. No.AMARH826640/DKD1Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826640/DKD3Specific gravity hydrometerSpecific gravity hydrometer51116 H - 621,250 - 1,300 °SG0,005 °SG330 mmPřihlásit se pro dostupnost1 * 1 KSSpecifikace:
More
Specifications
Cat. No.AMARH826640/DKD3Přesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C)
With factory test certificate
Informace o produktu
- SekvenceModelRozsahDěleníTeplotní rozsahDélkaDostupnostPrice
- AMARH826786WCSpecific gravity hydrometerSpecific gravity hydrometer50105 H - 620,800 - 0,850 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 150,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826786WCPřesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C) - AMARH826788WCSpecific gravity hydrometerSpecific gravity hydrometer51106 H - 620,850 - 0,900 °SG0,001 °SG260 mmPřihlásit se pro dostupnost1 * 1 KS1 150,00 KčSpecifikace:
More
Specifications
Cat. No.AMARH826788WCPřesnost±1 scale divisionReferenční teplota60/60 °F (except model AMARH826764-FL: 15 °C)