Specimen clamps for rotary microtomes
Catalog # LEIC14050238002
Supplier: LEICA MICROSYSTEMS
Specifications
- Popis:Holder for round specimens, 3 rings, with adapter with inserts
- Sekvence:01
- Bal.:1
Specifications
About this item
These clamps can be integrated into the fine directional, directional, or non-directional specimen holder fixtures.