HPLC columns, ZORBAX® Eclipse Amino Acid Analysis
HEWL963400-902
: Agilent
- Formát kolony:Analytical
- Ø vnitř.:4,60 mm
- Velikost částic:3,5 µm
- Délka:150 mm
- Sekvence:03
- Fáze:Eclipse Amino Acid Analysis
- Bal.:1
ZORBAX Eclipse AAA high efficiency column rapidly separates amino acids following an updated and improved protocol.
- High resolution and rapid analysis of 24 amino acids
- Uses well-known OPA and FMOC precolumn derivatisation chemistry
Total analysis from injection-to-injection can be achieved in as little as 8 min (7 min analysis time) on a 50 mm column, 14 min (9 min analysis time) on 75 mm length columns and 24 min (18 min analysis time) on the 150 mm column length.