0
0,00 Kč
Order Entry
Přihlásit se
/
Registrovat
Products
Services
Digital Solutions
Applications
Support
About
Expresní objednávka
Stav objednávky
Váš účet
Košík
Czech Republic
Expresní objednávka
Stav objednávky
Váš účet
Košík
ContactUsLinkComponent
Domů
Ovladače
Termostaty
Accessories for temperature control units
Accessories for temperature control units
Accessories for temperature control units
Catalog # JULA8980142
Supplier:
JULABO GmbH
Expand image
Accessories for temperature control units
Catalog # JULA8980142
Supplier:
JULABO GmbH
Specifications
Popis:
Y-adapter cable for connecting 1 dual PT100 sensor to 2 Lemosa sockets for LC temperature laboratory controller
Bal.:
1
Specifications
About this item
Product Details
Product Details