Capillary columns, OPTIMA® 17 MS
Supplier: MACHEREY-NAGEL
Certificates
About this item
Medium polar silarylene GC phase with low bleeding.
- Ideal for ion trap MS detectors
- Recommended for environmental analyses, ultra-trace analyses, EPA methods, pesticides, PCBs, food and drug analyses, also as reference column in combination with 1 MS or 5 MS columns
- Maximum temperature for isothermal operation 340 °C
- Maximum temperature for short isotherms in a temperature program 360 °C
The selectivity is analogue to 50% phenyl - 50% methylpolysiloxane phases.
USP code G3
Specifications
- Catalog No:
- MANA726165.60
- MANA726162.30
- MANA726162.60
- MANA726165.30
- Kód USP:
- G3
- G3
- G3
- G3