Orders LinkContactUsLinkComponent
Reading magnifiers, visoPOCKET
Reading magnifiers, visoPOCKET
Supplier:   ESCHENBACH OPTIK
Reading magnifiers, visoPOCKET
Supplier:  ESCHENBACH OPTIK
Certificates
Reading magnifiers, visoPOCKET
Reading magnifiers, visoPOCKET
3 Options Available Below
Product Details & Documents
Compact magnifiers with wide field of view and ultra-slim lenses.
  • Diffractive aspheric lens with modern and exquisite design
  • High quality leather available in three colours
  • Leather case protects from scratches
Sutiable for on-the-go use when small print needs magnifying swiftly and easily.
Product Family Options
Product Information
  • Popis
    Zvětšení
    Š×H
    Availability
    Price
  • Reading magnifier, red, visoPOCKET
    2,5×
    74×48 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 920,00 Kč
    920,00 Kč
    1 * 1 KS
       920,00 Kč
  • Reading magnifier, brown, visoPOCKET
    2,5×
    74×48 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 920,00 Kč
    920,00 Kč
    1 * 1 KS
       920,00 Kč
  • Reading magnifier, black, visoPOCKET
    2,5×
    74×48 mm
    Log in for availability
    Log in for availability
    1 * 1 KS
    1 * 1 KS - 920,00 Kč
    920,00 Kč
    1 * 1 KS
       920,00 Kč