Cell counter, Corning®
Product Details & Documents
This cell counter uses online image processing with a cloud-based machine learning algorithm.
- Fast and accurate results: Fits into the palm of a hand
- Cell count in less than 3 seconds
- Sophisticated deep neural network for cell detection with optimal accuracy – when Trypan blue is added, the system can also detect cell viability
Connect to a Windows® 10 computer or tablet and start the Cytosmart Cloud app. Place the counting chamber on stage, focus on the cells, and press the counting function.
The simplicity of the device allows anyone working in the lab to easily count cells without the need for extensive training. This system utilises a reusable counting chamber which is included, or a customer supplied haemocytometer.
Certifications: CE marked. usTUV, CSA certified.
Ordering information: Includes a reusable counting chamber.
Documents
Analyzing 3D Cell Culture using the Corning® Cell Counter Cell Counter, Corning Corning® Cell Counter Autofocus Functionality for Mammalian Cells Counts
Application Notes
Corning® Cell Counter Demo Video Corning® Cell Counter Introduction Trypan Blue Dilution Calculation and Histogram Gating for Corning® Cell Counter
Videos
TDS Corning® Cell Counter
Datasheet
Specifications
- Popis:Cell counter