Organic reference standard, 4-chloro-5-sulphamoylanthranilic acid, BP CRS
PROOBP074
:
- Bal.:25 mg
- Standard:Reference standard
- Čí. CAS:3086-91-7
- Popis:Chloro-5-sulphamoylanthranilic acid
- Vzorec:C₇H₇ClN₂O₄S
- Popis:Chloro-5-sulphamoylanthranilic acid