Organic reference standard, bis(2-propylheptyl) phthalate
Catalog # PROOTRCB523500-100
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Bal.:100 mg
- Standard:Reference standard
- Čí. CAS:53306-54-0
- Popis:Bis(2-propylheptyl) phthalate
- Vzorec:C₂₈H₄₆O₄
- Popis:Bis(2-propylheptyl) phthalate