Stem transfection reagents, DreamFect™ Stem
Dodavatel: OZ BIOSCIENCES
Certifikáty
O tomto produktu
DreamFect™ Stem is a powerful transfection reagent allowing multipotent stem cell transfection with high efficiency and very low toxicity. Its specific composition based on TEE-technology allows transfecting embryonic and multipotent stem cells in the presence of serum, maintaining their undifferentiated stage and their capacity to differentiate.
- High transfection efficiency for multipotent stem cells
- Minimised toxicity due to reagent biodegradability and low DNA amount required
- Cell phenotype and differentiation potential are not affected
- Serum compatible
- Simple, ready to use and rapid
Store at –20 °C.
Delivery: Shipped at room temperature.
Specifikace
- Specifické buňky:Stem cells
- DNA:Ano
- Vysoce výkonný skrínyng:Ano