DNA transfection reagent, DreamFect™ Gold
Dodavatel: OZ BIOSCIENCES
Certifikáty
O tomto produktu
DreamFect™ Gold transfection reagent allows transfecting all types of nucleic acids with a very high efficiency. Based on Tee-technology (Triggered Endosomal Escape), DreamFect™ Gold delivers a large quantity of nucleic acids leading to higher protein expression compared to other transfection reagents. It is fully and does not interfere with cellular mechanisms. Consequently, high cell viability is maintained in every experiment and any potential secondary effect is avoided.
- High efficiency
- Higher protein expression
- Superior protein expression level
- Avoid secondary effects
- Serum compatible
- Simple, rapid and easy to use
Store at –20 °C.
Delivery: Shipped at room temperature.
Specifikace
- DNA:Ano
- mRNA:Ano
- Oligos:Ano
- Vysoce výkonný skrínyng:Ano
- siRNA:Ano
Specifikace
Související informace
Frequently Bought Together