Order Entry
Czech Republic
ContactUsLinkComponent
Silikagel Rubin drying bags, crystalline 1.0-3.0 mm, with indicator 1/3 TE ( ~10 g/bag), Fluka™
Silikagel Rubin drying bags, crystalline 1.0-3.0 mm, with indicator 1/3 TE ( ~10 g/bag), Fluka™
Catalog # 80148-1KG
CAS Number:  
Silikagel Rubin drying bags, crystalline 1.0-3.0 mm, with indicator 1/3 TE ( ~10 g/bag), Fluka™
Catalog # 80148-1KG
CAS Number:  

Specifications

  • Typ balení:
    Metal can
  • Bal.:
    1 kg
  • Sekvence:
    4024

Specifications

Specification Test Results

  • Appearance of the substance:
    complying
  • Sieve analysis 1-3 mm:
    Min. 95 %
  • Water absorbing capacity:
    Min. 20 %
  • Suitability as indicator:
    complying

Specification Test Results

About this item

 :
 :60.08 g/mol
Boiling Pt:2,230 °C
Density:2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-545-4
CAS Number:112926-00-8
 :63231-67-4
Formula:SiO₂
MDL Number:MFCD00011232