1536-well microplates, cyclo olefin
Katalogové číslo GREI792870-906
Dodavatel: Greiner Bio-One
Specifikace
- Tvar:F-Bottom
- Balené:15/ bag
- Doporučený pracovní objem:3 - 10 µl
- Sterilní:-
- Barva:Průhledná
- Environmentally Preferable:Low Manufacturing Impact
- Certifikát ochrany životního prostředí:ISO 14001 (241002); ISO 50001 (241002)
- Bal.:60
- Objem jamky:12,6 µl
Specifikace
O tomto produktu
Clear, 1536-well cycloolefin storage plates (with optimised geometry for low evaporation) for acoustic liquid handling.
- Free from DNase, RNase, and human DNA
- Groove around the plate edge matches cycloolefin plate lid (not included) - designed to reduce evaporation and minimise edge effects
Acoustic liquid dispensing offers the advantage of pipetting small volumes very precisely and with minimal disturbance without the risk of cross-contamination. The process involves the transferral of DMSO-containing solutions in the nanoliter to picoliter range from a starting plate into a target plate by means of ultrasound.
Certifications: Manufactured under DIN ISO 9001 guidelines.
Ordering information: Plates are supplied without lids. Accessory lid is sold separately.