Iodide Iodate concentrate for dilution to 1/128 mol (1/64 N) in aqueous solution, ConvoL NORMADOSE® volumetric solution
Catalog # 84103.600
Supplier: VWR Chemicals
CAS Number:
Specifications
- Typ balení:Plastic ampoule
- Bal.:50 mL
- Sekvence:2370
Specifications
Specification Test Results
- Titer (N):0.0155-0.0157
Specification Test Results
About this item
:
:Dose for preparation of 1 litre of solution.
Storage Temperature:Ambient