Octan sodný trihydrát ≥97%, TECHNICAL
Catalog # VWRS27641.460
Supplier: VWR Collection
CAS Number:
Specifications
- Bal.:25 kg
- Sekvence:4283
Specifications
Specification Test Results
- Assay:Min. 97 %
Specification Test Results
About this item
:
Merck Index:13,08642
Boiling Pt:120…123 °C (1013 hPa)
Melting Pt:58 °C
Density:1,4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:204-823-8
CAS Number:6131-90-4
Formula:H₃CCOONa·3H₂O
MDL Number:MFCD00071557