Voda, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Katalogové číslo VWRS10292MB
Supplier: VWR Collection
CAS Number:
Specifications
- Bal.:1.000 L
- Sekvence:5037
Specifications
Specification Test Results
- Substances reducing KMnO4:Passes test
- Conductivity (at filling):Max. 1 µS/cm
- pH (25°C):5.0 - 7.5
- Evaporation residue:Max. 1 ppm
- Ignition residue:Max. 1 ppm
- Silica:Max. 0.1 ppm
- Cl (Chloride):Max. 0.05 ppm
- F (Fluoride):Max. 0.05 ppm
- NH4 (Ammonium):Max. 0.2 ppm
- NO3 (Nitrate):Max. 0.01 ppm
- PO4 (Phosphate):Max. 0.01 ppm
- SO4 (Sulphate):Max. 0.1 ppm
- Ag (Silver):Max. 0.0004 ppm
- Al (Aluminium):Max. 0.002 ppm
- As (Arsenic):Max. 0.002 ppm
- Au (Gold):Max. 0.001 ppm
- B (Boron):Max. 0.005 ppm
- Ba (Barium):Max. 0.001 ppm
- Be (Beryllium):Max. 0.002 ppm
- Bi (Bismuth):Max. 0.001 ppm
- Ca (Calcium):Max. 0.05 ppm
- Cd (Cadmium):Max. 0.001 ppm
- Co (Cobalt):Max. 0.001 ppm
- Cr (Chromium):Max. 0.0004 ppm
- Cu (Copper):Max. 0.0004 ppm
- Fe (Iron):Max. 0.001 ppm
- Ga (Gallium):Max. 0.004 ppm
- Ge (Germanium):Max. 0.001 ppm
- In (Indium):Max. 0.001 ppm
- K (Potassium):Max. 0.05 ppm
- Li (Lithium):Max. 0.002 ppm
- Mg (Magnesium):Max. 0.005 ppm
- Mn (Manganese):Max. 0.0004 ppm
- Mo (Molybdenum):Max. 0.002 ppm
- Na (Sodium):Max. 0.1 ppm
- Ni (Nickel):Max. 0.0004 ppm
- Pb (Lead):Max. 0.001 ppm
- Pd (Palladium):Max. 0.008 ppm
- Pt (Platinum):Max. 0.001 ppm
- Sb (Antimony):Max. 0.001 ppm
- Sn (Tin):Max. 0.001 ppm
- Sr (Strontium):Max. 0.0004 ppm
- Ti (Titanium):Max. 0.001 ppm
- Tl (Thallium):Max. 0.00005 ppm
- V (Vanadium):Max. 0.001 ppm
- Zn (Zinc):Max. 0.004 ppm
- Zr (Zirconium):Max. 0.001 ppm
- ISO Standard Water Grade 3:Passes test
Specification Test Results
About this item
:
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332