Foam cleaner, TASKI Sprint Multi E4c
Catalog # DIVY7513060
Supplier: Diversey
Specifications
- Kapacita:12×500 ml
- Popis:Foam cleaner, TASKI Sprint Multi E4c
- Bal.:12 Bottle
Specifications
About this item
Aerosol foam cleaner for all water-resistant hard surfaces. Ideal for vertical surfaces, the light foam sticks to the surface and ensures a long-term action.
- Good emulsifying ability
- Foam, long-lasting effect
- Ready for use
- Effectively removes grease and stains
- Easy to apply via aerosol
This cleaner effectively removes fat, fingerprints, coffee stains, ink and other types of stains. It cleans without leaving any streaks on matte surfaces.
Certifications: The surfactants used in this product are in accordance with EC Directives 73/404 / EEC and 73/405 / EEC and their amendments.