Chart recorder pens
Pens, long-nib, red, for use with chart recorder REC-112
Katalogové číslo 18-1004-23
Dodavatel: Cytiva
Přejít na:
Product Details & Documents
Pens for use with paper chart recorders.
- Response time of 0,2 s (2,55 to 97,5% FSD)
- Fibre tip
Available in blue or red.
Documents
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Další podrobnosti o produktu
Specifikace
- Popis:Pens, long-nib, red, for use with chart recorder REC-112