HPLC columns, Cogent HPS™ C8
Supplier: MICROSOLV
Certificates
About this item
Cogent HPS™ C8 columns are high-purity type-B, silica-based HPLC columns and have narrow particle distribution making them extremely high-quality.
- Double endcapped
- Features monofunctional bonding which provides greater stability and reproducibility
These type-B silica columns are made with extremely low metal content and are excellent choices for all types of analytes including bases, neutrals and acids by reversed phase or normal phase.
Specifications
- Catalog No:
- MICS75008-SP50
- MICS75008-SP100
- MICS75008-P50
- MICS75008-SP250
- MICS75008-SP150
- MICS75008-P100
- MICS75008-15P
- MICS75008-25P
- MICS75008-P250
- MICS75008-P150
- MICS75008-15P-2
- MICS75008-10P
- MICS75008-10P-2
- MICS75008-05P-2
- Naplnění uhlíkem:
- 11,5%
- 11,5%
- 11,5%
- 11,5%
- 11,5%
- 11,5%
- 11,5%
- 11,5%
- 11,5%
- 11,5%
- 11,5%
- 11,5%
- 11,5%
- 11,5%
- S navazanou skupinou:
- Double
- Double
- Double
- Double
- Double
- Double
- Double
- Double
- Double
- Double
- Double
- Double
- Double
- Double
- Rozsah pH:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- Povrchová oblast:
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- Teplotní limit:
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C
- 60 °C