Burettes, straight PTFE stopcock, class A
Catalog # HACH2636540
Supplier: Hach
Specifications
- Kapacita:25 ml
- Sekvence:01
- Dělení:0,1 ml
- Bal.:1
Specifications
About this item
Borosilicate glass, class A.
Clear glass straight bore stopcock with PTFE key.