Industrial gloves, AlphaTec® 58-735
Supplier: Ansell
Certificates
About this item
These gauntlet style gloves with INTERCEPT™ Cut Resistance Technology yarn and nitrile coating are used for chemical and cut protection.
- Nitrile barrier provides advanced chemical protection from many frequently used chemicals
- INTERCEPT™ Cut Resistance Technology provides ISO C cut protection
- ANSELL GRIP™ Technology for the handling of wet or oily parts providing enhanced dexterity, grip and comfort
Ideal for chemical blending and sampling, handling materials with sharp edges, laboratory work, metal working with corrosives, tools and parts cleaning.
Certifications: EN 388:2016 (4X32C); EN 374-5:2016; EN ISO 374-1:2016 Type A (AJKLMNOPT); Cat. III
Food contact approved
Food contact approved
Specifications
- Catalog No:
- 112-5164
- 112-5163
- 112-5162
- 112-5161
- 112-5165
- Model:
- 58-735
- 58-735
- 58-735
- 58-735
- 58-735
- Obouruké/jednoruké:
- Jednoruké
- Jednoruké
- Jednoruké
- Jednoruké
- Jednoruké
- Jednorázové/znovupoužitelné:
- opakovatelně použitelné
- opakovatelně použitelné
- opakovatelně použitelné
- opakovatelně použitelné
- opakovatelně použitelné
- Latex/bez latexu:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Materiál vložky:
- Intercept™ yarn
- Intercept™ yarn
- Intercept™ yarn
- Intercept™ yarn
- Intercept™ yarn
- Provedení:
- Sandy
- Sandy
- Sandy
- Sandy
- Sandy
- Rozchod:
- 18
- 18
- 18
- 18
- 18
- Typ manžety:
- Rukavice
- Rukavice
- Rukavice
- Rukavice
- Rukavice
- Norma EN:
- EN 388:2016 (4X32C); EN 374-5:2016; EN ISO 374-1:2016 Type A (AJKLMNOPT)
- EN 388:2016 (4X32C); EN 374-5:2016; EN ISO 374-1:2016 Type A (AJKLMNOPT)
- EN 388:2016 (4X32C); EN 374-5:2016; EN ISO 374-1:2016 Type A (AJKLMNOPT)
- EN 388:2016 (4X32C); EN 374-5:2016; EN ISO 374-1:2016 Type A (AJKLMNOPT)
- EN 388:2016 (4X32C); EN 374-5:2016; EN ISO 374-1:2016 Type A (AJKLMNOPT)
- Antistatický:
- +
- +
- +
- +
- +
- Schváleno pro styk s potravinami:
- +
- +
- +
- +
- +