Glass cleaner, Suma Rapid D6
Catalog # KINGLE511964
Supplier: KING DISPOSABLES
Specifications
- Popis:Glass cleaner, D6
- Typ balení:Spray bottle
- Balené:1×750 ml
- Bal.:1
Specifications
About this item
Suma Rapid D6 is a quick drying glass cleaner for use on windows, mirrors, display cabinets, tiles and other glazed ceramics.
- Easy spray-on, wipe-off action
- Quick drying with no residues
- Leaves glass sparkling with no streaks and smears
Certifications: Conforms to SABS 1828 'Cleaning Chemicals for use in the food industry'.