Chlorid železitý, anhydrous for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7705-08-0
:
Formula:FeCl₃
MDL Number:MFCD00011005
Merck Index:13,04048
Boiling Pt:315 °C (1013 hPa)
Melting Pt:260 °C
Density:2,8 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-729-4
ADR:8,III
:M-Clarity™ quality level = MQ200
UN:1773
:GHS05
:GHS07
Related Information
Certificates
Search by Lot Numbermsds_803945
SDS
ProductPage_803945 ProductPage_803945
Product specification
Frequently Bought Together