Přeskočit na Patička
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fluorescein dvojsodná sůl
Fluorescein dvojsodná sůl, pure for hydrology
Fluorescein dvojsodná sůl, pure for hydrology
Fluorescein dvojsodná sůl, pure for hydrology
Catalog # 720-1067
Supplier:
RAL
CAS Number:
Certificates
Expand image
Fluorescein dvojsodná sůl, pure for hydrology
Catalog # 720-1067
Supplier:
RAL
CAS Number:
Certificates
Specifications
Bal.:
1 kg
Sekvence:
4024
Specifications
About this item
Product Details
Documentation
:
CAS Number:
518-47-8
Formula:
C₂₀H₁₀Na₂O₅
MDL Number:
MFCD00167039
Product Details
Documentation