Titanium(IV) isopropoxide, Sigma-Aldrich®
Catalog # SIAL687502-25G
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:25 g
- Sekvence:3308
Specifications
About this item
:
Merck Index:12,09614
Boiling Pt:240 °C (1013 hPa)
Melting Pt:18…20 °C
Density:0,9711 g/cm³ (20 °C)
Flash Pt:51 °C
EINECS:208-909-6
UN:2413
ADR:3,III
:M-Clarity™ quality level = MQ100
CAS Number:546-68-9
Formula:C₁₂H₂₈O₄Ti
MDL Number:MFCD00008871