Orders LinkContactUsLinkComponent
Mop wipe systems, VertiKlean® MAX®
Mop wipe systems, VertiKlean® MAX®
Dodavatel:   CONTEC
Mop wipe systems, VertiKlean® MAX®
Dodavatel:  CONTEC
Certifikáty
Mop wipe systems, VertiKlean® MAX®
Mop wipe systems, VertiKlean® MAX®
3 Dostupných variant níže
Product Details & Documents
The VertiKlean® MAX Mop Wipe Systems offer a quick and easy solution to mopping floors, walls, and other flat surfaces in any controlled environment. These versatile systems work with a variety of mop wipe covers, enabling the user to customize the cleaning task.
  • Stainless steel frame is chemical resistant
  • Easy to attach and remove mop covers
  • Variety of fabric options available
  • Removes contaminants easily from the environment
  • Slim profile allows easy access to confined spaces for efficient cleaning and disinfection
  • Thin, lightweight design makes it easy to clean floors, walls, and ceilings
These can be sterilised by most common methods including gamma irradiation, ethylene oxide, or autoclave (recommended cycle is 121 °C for 30 minutes). Pre-saturated or dry wipes are available to eliminate the need for a bucket and wringer.
Dry and pre-saturated single-use wipes fit over the VertiKlean® MAX foam mop head and QuickConnect frame to remove contamination such as powder, dirt, and dust from floors, walls, and ceilings. The VertiKlean® MAX Mop Wipe Systems provide excellent surface contact and conformity. It can be used to apply disinfectant solutions to surfaces when used with a bucket and wringer. Most importantly, when used for disinfection, the mop wipe covers entrap microorganisms, enabling them to be removed from the environment with each change of wipe cover.
Product Family Options
Informace o produktu
  • Sekvence
    Barva
    D׊
    Balené
    Dostupnost
    Price
  • 115-3201
    Polyester quilted mop wipes, Quiltec®
    Polyester quilted mop wipes, Quiltec®
    01
    Bílá
    250×420 mm
    20 wipes/bag; 12 bags/case
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 240 KS
    1 * 240 KS - 18 360,00 Kč
    18 360,00 Kč
    1 * 240 KS
       18 360,00 Kč
  • 115-3202
    Polyester quilted mop wipes, presaturated with 70% IPA and 30% DI water, Quiltec®
    Polyester quilted mop wipes, presaturated with 70% IPA and 30% DI water, Quiltec®
    02
    Bílá
    250×420 mm
    20 wipes/bag; 9 bags/case
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 180 KS
    1 * 180 KS - 30 060,00 Kč
    30 060,00 Kč
    1 * 180 KS
       30 060,00 Kč
  • 115-1861
    Mop jednorázový, MicroGenesis™
    Mop jednorázový, MicroGenesis™
    03
    Bílá
    250×460 mm
    100 ks/sáček; 4 sáčků/balení
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 400 KS
    1 * 400 KS - 20 380,00 Kč
    20 380,00 Kč
    1 * 400 KS
       20 380,00 Kč