
Chlorid amonný, Sigma-Aldrich®
Catalog # SIAL09724-1KG
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:1 kg
- Sekvence:4024
Specifications
About this item
:
Merck Index:13,00510
Melting Pt:335 °C
Density:1,5256 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:235-186-4
:GHS07
Storage Temperature:Ambient
CAS Number:12125-02-9
:MFCD00143604
Formula:NH₄Cl
MDL Number:MFCD00011420