Order Entry
Czech Republic
ContactUsLinkComponent
Human thrombin (from blood)
Human thrombin (from blood)
Catalog # CAYM13188-1
Supplier:  Cayman Chemical
CAS Number:  
Human thrombin (from blood)
Catalog # CAYM13188-1
Supplier:  Cayman Chemical
CAS Number:  

Specifications

  • Bal.:
    1 KU
  • Čí. CAS:
    9002-04-4
  • Zdroj:
    Blood
  • Druhy:
    Human
  • Sekvence:
    02
  • Podmínky uskladnění:
    –80 °C
  • Název enzymu:
    Thrombin
  • Aktivita enzymu:
    ≥2000 NIH units/mg
  • Synonyma enzymů:
    Activated factor II [IIa]
  • Molekulární hmotnost:
    38 kDa

Specifications

About this item

Thrombin is the last enzyme in the clotting cascade functioning to cleave fibrinogen to fibrin and potentiates the procoagulation process by activating factors V, VIII, XI, and XIII.

Thrombin aids the anticoagulant process by activating protein C. Activated protein C inhibits activated factors V and VIII ultimately down-regulating further thrombin production. Thrombin is often used as a potent platelet activator in vitro. Platelet responses to thrombin are mediated through protease-activated receptors (PARs). Thrombin-mediated signaling regulates a diverse range of metabolic processes including activation of phospholipases in platelets, endothelial cells, and cardiac myocytes.

Formulated as 5 mM sodium citrate, pH 6,5, containing 0,01% PEG and 20 mM sodium chloride.