ContactUsLinkComponent
Safety scissors, SECUMAX 564
Safety scissors, SECUMAX 564
Katalogové číslo MARO564001.00
Dodavatel:  MARTOR ARGENTAX
undefined
Safety scissors, SECUMAX 564
Katalogové číslo MARO564001.00
Dodavatel:  MARTOR ARGENTAX
Specifikace
  • Type:
    Safety scissor, SECUMAX 564, 80 mm size edge, 218×12×82 mm
  • Bal.:
    1
Specifikace
O tomto produktu
The SECUMAX 564 is characterised by its thumb opening and its extra-long edge, which also makes an ideal support surface. These safety scissors enable you to completely control your work.
  • Sharp, but not dangerous Non-rusting stainless steel Extra-long cutting edges Robust, fiber-glass reinforced plastic handle
Highest safety guaranteed by the special sharpening of the cutting blades and the rounded tips.