Order Entry
Czech Republic
ContactUsLinkComponent
Tetramethyltin for synthesis, Sigma-Aldrich®
Flammable
Toxic
Environment
Tetramethyltin for synthesis, Sigma-Aldrich®
Catalog # 8.08706.0010
Supplier:  Merck
CAS Number:  
Flammable
Toxic
Environment
Tetramethyltin for synthesis, Sigma-Aldrich®
Catalog # 8.08706.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Typ balení:
    Glass bottle
  • Bal.:
    10 mL
  • Sekvence:
    2275

Specifications

Specification Test Results

  • Solubility:
    (20 °C) practically insoluble
  • Melting Point:
    -54 °C
  • Molar Mass:
    178.82 g/mol
  • Boiling Point:
    74 - 75 °C (1013 hPa)
  • Flash Point:
    -12 °C
  • Refractive Index:
    1.441 (20 °C, 589 nm)
  • Density:
    1.29 g/cm3 (20 °C)

Specification Test Results

About this item

 :
Boiling Pt:75 °C (1013 hPa)
Melting Pt:–54 °C
Density:1,29 g/cm³ (20 °C)
UN:1992
ADR:3 (6.1),II
 :M-Clarity™ quality level = MQ200
Flash Pt:–12 °C
EINECS:209-833-6
 :GHS02
 :GHS06
 :GHS09
Storage Temperature:Ambient
CAS Number:594-27-4
Formula:C₄H₁₂Sn
MDL Number:MFCD00008278