Uhličitan vápenatý, Sigma-Aldrich®
Catalog # SIAL21069-1KG
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:1 kg
- Sekvence:4024
Specifications
About this item
:
Merck Index:13,01658
Melting Pt:825 °C
Density:2,71 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:207-439-9
CAS Number:471-34-1
Formula:CaCO₃
MDL Number:MFCD00010906