Uhličitan vápenatý, EMPROVE® ESSENTIAL Ph. Eur., BP, USP, JP, FCC (max. 0.002% Fe), precipitated, SAFC®
Catalog # 1.02064.9029
Supplier: MERCK PRODUCTION CHEMICALS
CAS Number:
Specifications
- Bal.:25 kg
- Sekvence:4283
Specifications
About this item
:
Merck Index:13,01658
Melting Pt:825 °C
Density:2,71 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:207-439-9
CAS Number:471-34-1
Formula:CaCO₃
MDL Number:MFCD00010906