1-Methylpiperazine, Supelco®
Catalog # 14549-5ML-F
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle
- Bal.:5 mL
- Sekvence:2210
Specifications
About this item
:
Boiling Pt:138 °C (1013 hPa)
Melting Pt:–6 °C
Density:0,9085 g/cm³ (20 °C)
Storage Temperature:Ambient
UN:2734
ADR:8,II
:M-Clarity™ quality level = MQ100
Flash Pt:34 °C
EINECS:203-639-5
CAS Number:109-01-3
Formula:C₅H₁₂N₂
MDL Number:MFCD00005966