ContactUsLinkComponent
2-Methylhydroquinone for synthesis, Sigma-Aldrich®
Toxic
2-Methylhydroquinone for synthesis, Sigma-Aldrich®
Catalog # 8.20801.0100
Supplier:  Merck
CAS Number:  
Toxic
2-Methylhydroquinone for synthesis, Sigma-Aldrich®
Catalog # 8.20801.0100
Supplier:  Merck
CAS Number:  
Specifications
  • Typ balení:
    Glass bottle
  • Bal.:
    100 g
  • Sekvence:
    3401
Specifications
Specification Test Results
  • Solubility:
    77 g/l (25 °C)
  • Melting Point:
    124 - 127 °C
  • Molar Mass:
    124.14 g/mol
  • Boiling Point:
    285 °C
  • Flash Point:
    172 °C
  • Ignition Point:
    468 °C
Specification Test Results
About this item
 :
Boiling Pt:285 °C (1013 hPa)
Melting Pt:123…126 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
 :M-Clarity™ quality level = MQ200
EINECS:202-443-7
 :GHS06
CAS Number:95-71-6
Formula:C₇H₈O₂
MDL Number:MFCD00002345