2-Nitro-p-cresol
Catalog # EHERC15110800
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:0,1 g
- Sekvence:3020
Specifications
About this item
:
Boiling Pt:234 °C (1013 hPa)
Melting Pt:31 °C
Density:1,2399 g/cm³ (39 °C)
EINECS:204-315-6
UN:2446
ADR:6.1,III
:GHS07
CAS Number:119-33-5
Formula:C₇H₇NO₃
MDL Number:MFCD00007120