Apparatus with flange for the determination of Arsenic
Catalog # NEUB23900041002920
Supplier: NEUBERT VOLUME GLASSWAERE
Specifications
- Popis:Apparatus, complete
- Bal.:1
Specifications
About this item
Borosilicate glass 3,3.
Apparatus for the estimation of Arsenic. Complete including two springs, erlenmeyer flasks 100 ml, graduated, ST 29/32, tube with ST 29/32 and flange.
Certifications: DAB 10 (Pharmeuropa 28.2 ; Ph.Eur 2.4.2 Method A )